VWR® Professional Magnetic Stirrers
Supplier: VWR Collection
444-0616EA 1520 EUR
444-0616 444-0611 444-0612 444-0613 444-0614
VWR® Professional Magnetic Stirrers
Stirrers Magnetic Stirrers
VWR® Professional magnetic stirrers are designed for applications that require exceptional accuracy, stability and repeatability. Stirring function, with continuous duty motor and powerful magnet, maintains set speed even under changing load or viscosity. Touch pad controls with easy to read, independent LED displays for speed, and time, allow the operator to view all settings at once.
  • Microprocessor control
  • Separate digital displays for speed and time. Display will show last used settings, even after power has been turned off
  • Chemically resistant housing
More Product Information