VWR® 3D Rotator Waver
Supplier: VWR Collection
444-0759EA 1810 EUR
444-0759 444-0760 444-0761
VWR® 3D Rotator Waver
Shakers Rotating/Rocking Shakers
The low profile 3-D rotator waver provides precise speed control and a smooth, low foaming, three dimensional, wave motion. The cast aluminium base offers durability and added stability, the spill resistant design channels fluids away from internal components. The microprocessor control provides electronic tilt angle adjustment which allows user to easily and accurately adjust waving angle from 0 to 20° while unit is operating. Precise speed control provides smooth, low speed waving motion down to 1 min⁻¹.
  • Electronic tilt adjustment from 0 to 20° while unit is operating
  • LED display
More Product Information