Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
504-0233-00 146-3011-00 0K-0500-901 580-2013-00 146-6005-00 SI-0570 SI-0511 503-0280-00 503-0279-00 503-0277-00 503-0278-00 568-0001-00 504-0039-00 503-0283-00 504-0234-00 504-0235-00 503-0282-00 SI-0510 0K-0500-902 0K-0513-900 SI-0513 SI-V512 SI-H524 SI-H506 SI-H512 SI-V525 SI-0564 SI-0565 SI-V506 SI-V524 SI-V203 SI-V209 0K-0236-408 SI-V206 504-0232-00 SI-HP524
444-5904EA 222 EUR
444-5904 444-5901 444-5902 444-5903 444-5905 444-9161 444-9162 444-5911 444-5912 444-5913 444-5914 444-5915 444-5916 444-5917 444-5921 444-5919 444-0060 444-0061 444-0443 SCII0K-0513-900 SCIISI-0513 444-1050 444-1045 444-1043 444-1044 444-1052 444-0724 444-0775 444-1049 444-1051 444-1046 444-1048 SCII0K-0236-408 444-1047 SCII504-0232-00 SCIISI-HP524
Accessories for vortex mixers Vortex-Genie®
Shakers Vortex Shakers
* Multiple sample starter set consists of a 152 mm platform, a foam insert for 60 microtubes and a foam insert for one standard microplate
** Large sample set is ideal for mixing beakers, flasks and large tubes. The set consists of a recessed platform, two elastic bands, a blank foam insert, a foam insert for 4×29 to 37 mm tubes, a foam insert for 8×14 to 29 mm tubes and a foam insert for 12×9 to 16 mm tubes
*** Small ampoule/tube attachment enables vigorous end-to-end mixing for up to four 15 to 17 mm Ø tubes or ampoules
**** Large ampoule/More Product Information