Reciprocating shakers, KS 15 B and KS 15 B control
Supplier: Edmund Bühler GmbH
6173000 6172000
444-9220EA 2140 EUR
444-9220 444-9222
Reciprocating shakers, KS 15 B and KS 15 B control
Shakers Reciprocating Shakers
Reciprocating shakers, which are suitable for continuous or timer operation. The optional universal tray accepts a maximum of 5×1000 ml or 9×500 ml flask clamps; Combifix rack system accommodates 4×1000 or 6×500 ml Erlenmeyer flasks. Variety of different vessels can be accommodated by choosing from the range of optional attachments.
  • KS 15 B has analogue control, KS 15 B control has digital display and a new programmable control with extended functionality
  • Continuous operation or timer operation from 0 to 120 min (KS 15 B), and continuous
More Product Information