1,4-Dioxane 99% stabilised, AcroSeal® for biochemistry

Supplier: ACROS ORGANICS
Danger

447060010 447060025
447060010.EA 108 EUR
447060010. 447060025.
1,4-Dioxane 99% stabilised, AcroSeal® for biochemistry
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1.034 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=64c930c52bed4490a4dd38741bf8cb01a966ce0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.1 %
Heavy metals ≤5 ppm
GC ≥98.5 %
UV at 275 nm A: ≤0.1
Color scale ≤10 APHA
Stabilizer 2.5 ppm BHT
Residue after evaporation ≤0.01 %
Peroxides ≤0.005 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR