Aluminium oxide, γ-phase ≥99%, nanopowder

Supplier: ALFA AESAR

44757.14 44757.22 44757.36
44757.14EA 68.5 EUR
44757.14 44757.22 44757.36
Aluminium oxide, γ-phase ≥99%, nanopowder
Aluminium oxide, γ-phase

Purity: 99+% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 20nm APS Powder, S.A. 100-200m2/g Appearance: White Odour: Odourless Merck Reference: 14,356 MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR