N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®

Supplier: ACROS ORGANICS
Danger

448381000
448381000.EA 86.8 EUR
448381000.
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
N,N-Dimethylformamide

Package of 4x25ML bottles

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.945 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
UN: 2265
ADR: 3 III
Merck Index: 13,03269
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=fb9c58451426a2acf92f6525238f8198a167f088&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.75 %
Residue after evaporation ≤0.001 %
Filt.over 0.2 micron PFTE Filtered
Acidity (CH3COOH) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR