Optical windows
Supplier: Alfa Aesar
45387.KS 45386.KS 45380.KS 45560.KS 45834.KS 46107.KS 45717.KS 45628.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
45387.KSEA 319 EUR
45387.KS 45386.KS 45380.KS 45560.KS 45834.KS 46107.KS 45717.KS 45628.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
Optical windows
Optical Windows
These optical windows are ideally stored at ambient temperatures.
  • Clear aperture 85%
  • Surface finish 60/40 or 40/20