Acetic acid ≥99,0% ≥99.0%, TraceSELECT™ for trace analysis, Fluka™

Supplier: HONEYWELL
Danger

45727-500ML 45727-1L 45727-2.5L
45727-500MLEA 164 EUR
45727-500ML 45727-1L 45727-2.5L
Acetic acid ≥99,0% ≥99.0%, TraceSELECT™ for trace analysis, Fluka™
Acetic acid

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: H₃CCOOH
Boiling Pt: 118 °C (1,013 hPa)
Melting Pt: 16 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=031896546f0eb78526a541f0af4636e3991e9f8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

assay (HPLC) Min. 99 %
measurement uncertainty effective %
identity (NMR) complying
melting range 62 - 78 °C
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
loss on drying (105°C, 5h) 15 - 30 %
Calcium (Ca) Max. 20 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 2 ppb
Iron (Fe) Max. 10 ppb
Gallium (Ga) Max. 0.5 ppb
Germanium (Ge) Max. 0.5 ppb
Mercury (Hg) Max. 1 ppb
Indium (In) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 5 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 20 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 1 ppb
Zinc (Zn) Max. 2 ppb
Zirconium (Zr) Max. 0.5 ppb
Refractive Index n20/D 1.371 (lit.)
Explosion Limit 16 %, 92 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR