Baths and circulators, polycarbonate, CORIO™ C-BT and CD-BT series
Supplier: JULABO GmbH
9011305 9011309 9011319 9011327 9012305 9012319 9012327
461-0888EA 1520 EUR
461-0888 461-0889 461-0890 461-0891 461-0897 461-0898 461-0899
Baths and circulators, polycarbonate, CORIO™ C-BT and CD-BT series
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORIO C immersion circulator is the basic model, it is ideal for standard internal applications, the CORIO CD model is more advanced and can be used for temperature control of external applications if required. The jet nozzle allows continuous adjustment of the pump stream in the bath. The transparent polycarbonate baths are durable and enable the user to easily view items in the bath.
  • Precise temperature control
  • Bright, white, easy to read
More Product Information