Drying cabinets, standard and insulated
Supplier: GENLAB
DC250 DC500/F IWC100 IWC200 IWC425 IWC885 DC1000/F DC125
DC250EA 1840 EUR
DC250 DC500/F 466-0233 466-0234 466-0235 466-0236 466-0229 466-0230
Drying cabinets, standard and insulated
Ovens Drying Cabinets, Glassware
These Genlab drying cabinets are suitable for general purpose warming applications and drying glassware or instruments. Made from sheet steel finished in an easy to clean powder coated paint. Fitted with adjustable shelf runners, removable plated wire grid shelves and ventilation holes. Insulated models (IWC) give energy savings of over 30% compared to the non-insulated range (DC). Heated by Incoloy sheathed elements in the base of the unit. The 500 and 1000 litre models have fan assistance as standard. All units have toughened glass doors; 125 and 250 litre models have sliding doors and theMore Product Information