Cryogenic chest freezer, VIP Plus™, −150 °C, PHCbi
Supplier: PHC Corporation
MDF-C2156VAN
471-0731EA 31762.68 EUR
471-0731
Cryogenic chest freezer, VIP Plus™, −150 °C, PHCbi
Freezers Cryogenic Freezers
Ideal for long-term preservation of biologicals, blood components and various cells below the recrystallisation point. With specially designed compressor and cascade refrigeration system for a durable ultra-low temperature pull-down. Can hold up to 14000×2 ml tubes, CFC-free. Microprocessor control with LED for accurate temperature control and a data logging display.
  • VIP Plus™ Insulation (polyurethane/VIP 135 mm) provides 25% more storage without increasing the footprint
  • Built-in audible and visual alarms for temperature and power failure
  • Door handle with
More Product Information