Organic reference standards, Menaquinone (Vitamin K2), analytical standard, Supelco®
Supplier: SUPELCO
47774
47774.EA 81.6 EUR
47774.
Organic reference standards, Menaquinone (Vitamin K2), analytical standard, Supelco®
Standards Organic Standards
Menaquinone (K2), a vitamin K2 homologue with four isoprene units. It has been reported to enhance bone formation and plays a vital role in the production of blood coagulation factors.
Menaquinone may be used as a reference standard for the determination of menaquinone in human plasma using liquid chromatography with tandem mass spectrometry.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1457a92e9a289502c6373ff9bb77ca13a0bec94a&vid=1457a92e9a289502c6373ff9bb77ca13a0bec94a&prid=web&clid=SAPDAM