Liquid nitrogen, low level alarms for Dewars
Supplier: QUANTUM PRODUCTION
QLA100 QLA101
478-0474EA 864 EUR
478-0474 478-0475
Liquid nitrogen, low level alarms for Dewars
Level Sensors
Low level alarms, easily fitted to any type or size of low pressure liquid nitrogen refrigerator, cryostat, vessel or Dewar flask. It continuously monitors levels of LN₂ and gives visual and audible warnings when LN₂ levels fall below the level set by the sensor position.
  • Independent warning of sensor damage
  • Remote alarm connection to in-house monitoring systems
  • Sensor mounting kit required
  • Adjustable sensor position
  • 15 V power supply included
Ordering information: Purchase of alarm with probe assembly must be specified at the time of ordering DewarMore Product Information