Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
3710 3711 3712-11 3715 3774 3770 3731-11 3740 3741 3729 3734 3732 3735 3736 3737 3742 3743 3744 3745 3775 3778 3791 3792 3801 3850 3744YEL 3750 3730-11 3744PUR 3744GRE 3744RED 3744WHI 3741AMB 3742AMB 3790 3741PUR 3743GRE SP-3744BLK SP-3744ORG 3746 3740PUR 3743BLU 3740BLU 3740GRE 3740RED 3743RED 3748 3741BLU 3741GRE 3741RED 3741WHI 3741YEL 3744BLU 3743YEL 3743PUR 3743WHI 3745-WP-BR 3742AMB-BR 3744-BR 3743AMB-BR 3744BLU-BR 3744GRE-BR 3744PUR-BR 3744RED-BR 3741-BR 3741AMB-BR 3741BLU-BR 3741GRE-BR 3741PUR-BR 3741RED-BR 3741WHI-BR 3741YEL-BR 3742-BR 3745-BR 3744WHI-BR 3744YEL-BR 3745AMB-BR 3743AMB 3740WHI 3740YEL 3725
479-1015EA 532 EUR
479-1015 479-1016 479-1017 479-1020 479-1058 479-1057 479-1024 479-1030 479-1031 479-1022 479-1026 479-1025 479-1027 479-1028 479-1029 479-1036 479-1040 479-1042 479-1052 479-1059 479-1060 479-1062 479-1063 479-1065 479-1066 479-1051 479-1056 479-1023 479-1046 479-1045 479-1047 479-1048 479-1032 479-1037 479-1061 479-1033 479-1041 MATRSP-3744BLK MATRSP-3744ORG MATR3746 479-2021 479-2032 479-2019 479-2020 479-2022 479-2034 479-1194 479-1228 479-1229 479-1227 479-1230 479-1226 479-1337 479-2036 479-2033 479-2035 MATR3745-WP-BR MATR3742AMB-BR 479-2038 479-2196 479-2037 479-2039 479-2040 479-2041 479-2025 479-2023 479-2024 479-2026 479-2027 479-2028 479-2029 479-2030 479-2031 479-2044 479-2042 479-2043 MATR3745AMB-BR MATR3743AMB MATR3740WHI MATR3740YEL MATR3725
Storage tubes, 2D barcoded, Matrix™
Vials
Permanently attached, laser-etched 2D barcodes on the bottom of each tube allow you to scan and decode tubes at once without removing them from the rack and allow data to be associated with each tube.
  • The 8×12 format enables the filling of 8, 12 or 96 tubes at once
  • Tubes come in latch racks for added security and protection of samples
  • Built-in error correction ensures that barcodes will never be misread
  • 100% quality control checks that every code is unique and readable
  • Solid, one-piece construction enables storage down to cryogenic temperatures
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement, ACT Label NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label NAR Certificate of Retirement