VWR® Cryoboxes and Dividers
Supplier: VWR Collection
478-0249EA 0 EUR
478-0249 479-2216 479-2217 479-2219 479-2220 479-2221 479-2223 479-2224 479-2225 479-2226 479-2227 479-2228 479-2213 479-2215
VWR® Cryoboxes and Dividers
Boxes Cryoboxes
Cardboard, white.
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.