Autoclaves, LABOKLAV
Supplier: SHP STERILTECHNIK
210055001000V 210055001000H 210160001000V 210160001000H 210195001000H 210195001000V 210100001000V 210080001006V 210080001004V 210080001004H 210100001000H 210080001000V 210080001000H 210135001000V 210135001000H
481-0402EA 17760 EUR
481-0402 481-0401 481-0465 481-0464 481-0480 481-0481 481-0434 481-0430 481-0426 481-0425 481-0433 481-0418 481-0417 481-0449 481-0448
Autoclaves, LABOKLAV
Sterilisers Autoclaves (Steam Sterilisers)
A completely new concept allows the installation of the unit as either front or top loader. Deciding on horizontal or vertical is no longer the question with SHP autoclaves. No matter if installed horizontally or vertically, the units remain the same. Even at a later stage the unit can be changed from vertical to horizontal installation or vice versa without the need for big technical changes (contact Avantor for details). When set up as horizontal units they are even stackable. Stainless steel 316L chamber and housing.
  • Automatic feed water pump
  • Fully automatic closing system and
More Product Information