N,N-Dimethylformamide ≥99.9%, Biotechnology Grade, Burdick & Jackson™

Supplier: HONEYWELL
Danger

494488-1L 494488-2L
494488-1LEA 112 EUR
494488-1L 494488-2L
N,N-Dimethylformamide ≥99.9%, Biotechnology Grade, Burdick & Jackson™
N,N-Dimethylformamide

Beilstein Number: 605365

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 152…153 °C (1.013 hPa)
Melting Pt: –61 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 57.5 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=fb9c58451426a2acf92f6525238f8198a167f088&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.005 %
Amines (as dimethylamine) Max. 8 ppm
Titrable Acid Max. 0.0005 meq/g
Titrable Base Max. 0.003 meq/g
Absorbance at 400 nm Max. 0.01
Absorbance at 340 nm Max. 0.01
Absorbance at 310 nm Max. 0.05
Absorbance at 295 nm Max. 0.10
Absorbance at 275 nm Max. 0.30
Absorbance at 270 nm Max. 1.00
Refractive Index n20/D 1.430 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR