Disodium hydrogen phosphate

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ100

5.43838.0100 5.43838.0250
5.43838.0100EA 303 EUR
5.43838.0100 5.43838.0250
Disodium hydrogen phosphate
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
MDL Number: MFCD00003496
CAS Number: 7558-79-4
Merck Index: 12,08805

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR