Dialysis membrane tubing Spectra/Por® 6
Supplier: Spectrum Laboratories
132540 132542 132544 132550 132552 132554 132560 132562 132564 132566 132570 132572 132574 132576 132579 132580 132582 132584 132586 132590 132592 132594 132620 132625 132633 132636 132638 132640 132562T 132540T 132566T 132542T 132544T 132550T 132552T 132554T 132560T 132570T 132564T 132592T 132572T 132574T 132576T 132579T 132580T 132582T 132584T 132590T 132594T 132620T 132625T 132633T 132636T 132638T 132640T 132586T
734-0633EA 675 EUR
734-0633 734-0634 734-0635 734-0636 734-0637 734-0638 734-0639 734-0640 734-0641 734-0642 734-0643 734-0644 734-0645 734-0646 734-0647 734-0648 734-0649 734-0650 734-0651 734-0652 734-0653 734-0654 734-0656 734-0657 734-0658 734-0659 734-0660 734-0661 514-0922 514-0915 514-0924 514-0916 514-0917 514-0918 514-0919 514-0920 514-0921 514-0925 514-0923 514-0935 514-0926 514-0927 514-0928 514-0929 514-0930 514-0931 514-0932 514-0934 514-0936 514-0937 514-0938 514-0939 514-0940 514-0941 514-0942 514-0933
Dialysis membrane tubing Spectra/Por® 6
Tubing Dialysis Tubing
MWCO 1 to 50 kD, hydrophilic cellulose.
  • Transparent and flexible
This economical membrane is ideal for a broad range of applications in which there is a significant size difference between the MW species being separated; including sample prep, desalting, buffer exchange, pH change, protein purification. Offering a good chemical compatibility, Standard Grade RC can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols and some mild or dilute organics, including DMSO. Standard RC can tolerate pH 2 to 12 andMore Product Information