n-Heptane, Supelco®

Supplier: SUPELCO
Danger


M-Clarity™ quality level = MQ100

51730-50ML 51730-5ML
51730-50MLEA 389 EUR
51730-50ML 51730-5ML
n-Heptane, Supelco®
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98,4 °C (1013 hPa)
Melting Pt: –90,6 °C
Density: 0,6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8cfb5b8944c352d16a53912949129f7b7d2e855&vid=c8cfb5b8944c352d16a53912949129f7b7d2e855&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=8448ee44d4a93782bfa2900878099424dfa17053&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR