VWR® Mega Star 1.6/1.6R, Centrifuges, Bench Top, Ventilated/Refrigerated
Supplier: VWR Collection
521-2658EA 4920 EUR
521-2658 521-2659 521-2660 521-2661
VWR® Mega Star 1.6/1.6R, Centrifuges, Bench Top, Ventilated/Refrigerated
Centrifuges Benchtop Centrifuges
These versatile centrifuges are ideal for a range of routine applications in pharmaceutical and biotechnology industries, as well as for academic research. They offer good capacity with ergonomic design which facilitates work on a bench top.
  • Auto-Lock® III rotor system: Tool-free rotor exchange system enables quick rotor exchange; with just the push of a button users can quickly change rotors and easily access the rotor chamber for cleaning
  • Aerosol-tight ClickSeal® bucket sealing system: Glove friendly, one-handed open/close capability, no need for screw
More Product Information