Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
0030119.380 0030119.452 0030119.487 0030119.401 0030119.460 0030119.479 0030119.606
525-0671EA 164 EUR
525-0671 525-0743 525-0797 525-0794 525-0795 525-0796 525-1044
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Tubes Microcentrifuge Tubes
Eppendorf Tubes® with snap caps are an ideal choice when working with medium-sized sample volumes (0,5 to 5,0 ml). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25000×g), and sample recovery.
  • Designed for ease of use and user-friendly handling during sample preparation and processing
  • Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions
  • Specifically designed to enhance sample recovery, preserving the integrity of samples
  • Available in Eppendorf Quality, PCR Clean,
More Product Information