Pump tubing, Tygon®
Supplier: Heidolph Instruments GmbH & Co.KG
525-60013-00 525-60014-00 525-60015-00 525-60016-00 525-60022-00 525-60023-00 525-60024-00 525-60025-00 525-60026-00 525-60027-00 525-60028-00 525-62000-00 525-63000-00 525-64000-00 525-65000-00 525-66000-00 525-69000-00 525-70027-00 525-70028-00 525-73000-00 525-74000-00 525-76000-00 525-80027-00 525-80028-00 525-83000-00 525-84000-00 525-85000-00 525-86000-00 525-89000-00 525-72000-00
525.60013.00EA 29.3 EUR
525.60013.00 525.60014.00 525.60015.00 525.60016.00 525.60022.00 525.60023.00 525.60024.00 525.60025.00 525.60026.00 525.60027.00 525.60028.00 525.62000.00 525.63000.00 525.64000.00 525.65000.00 525.66000.00 525.69000.00 525.70027.00 525.70028.00 525.73000.00 525.74000.00 525.76000.00 525.80027.00 525.80028.00 525.83000.00 525.84000.00 525.85000.00 525.86000.00 525.89000.00 HEID525-72000-00
Pump tubing, Tygon®
Tubing Transfer Tubing
Thermoplastic soft PVC, transparent.
  • Non-toxic, non-oxidizing
  • Good resistance to acids, bases and inorganic media
  • Very low gas permeability, good performance life
For standard applications.
Certifications: FDA and GLP conform.
Order Now
SPECIFICATIONS
Environmentally Preferable