Overhead shaker, Reax 2
Supplier: Heidolph Instruments GmbH & Co.KG
541-21001-00 541-21001-06 541-21001-07 541-21009-00 541-21009-03 541-21009-04
541.21001.00EA 1620 EUR
541.21001.00 444-1455 444-1495 444-0984 444-1067 444-1068
Overhead shaker, Reax 2
Shakers Overhead Shakers
Overhead shaker with one dimensional swirling action, designed for homogeneous mixing, ideal for haematology, medical diagnostics, materials testing and biochemistry. Unit has an analogue dial for adjusting speed setting.
  • Designed for continuous use
  • Easy to change universal adapter means various vessels can be used
  • Ideal for use under controlled conditions, can be used at 0 to 40 °C (80% relative humidity), in an incubator or cold room
IP protection class according to DIN EN 60529: IP 21
Complies with DIN 38414 part 4 - Determination of effluent inMore Product Information