Glass tubings, DURAN® borosilicate glass
Supplier: Schott AG
1026996 1026245 1026089 1026972 1026108 1026247 1026103 1026258 1026969 1026872 1026964 1026120 1026963 1026965 1026244 1026993 1026871 1026979 1026267 1026985 1026115 1026980 1026255 1026253 1026998 1026977 1026967 1026966 1026264 1026989 1026252 1026974 1026248 1026870 1026251 1026249 1026970 1026109 1026988 1026114 1026968 1026257 1026262 1026984 1026987 1026990 1026256 1026263 1026111 1026994 1026119 1026973 1026978 1026268 1026975 1026084 1026266 1026246 1026992 1026254 1026995 1026265 1026983 1026991 1025260 1026261 1026672 1026962 1026971 1026976 1026981 1026982 1026986 1026085 1026091 1026096 1026107
130(3.0)EA 848 EUR
130(3.0) 52(3.5) 55(5.0) 70(2.2) 100(9.0) 54(3.5) 80(9.0) 75(3.2) 60(4.2) 50(3.5) SCOR1026964 135(7.0) 52(2.5) 56(3.5) 52(1.8) 120(7.0) 50(2.5) 90(3.5) 95(3.5) 100(5.0) 115(7.0) 90(5.0) 65(4.2) 65(2.2) 130(5.0) 80(3.5) 58(2.5) 58(1.8) 85(5.0) 110(5.0) 60(3.2) 70(5.0) 56(1.8) 50(1.8) 58(3.5) 56(2.5) 60(5.0) 105(3.0) 110(3.0) 115(3.0) 60(2.2) 75(2.2) 85(2.5) 100(3.5) 105(5.0) 115(5.0) 70(3.2) 85(3.5) 110(7.0) 120(9.0) 125(9.0) 70(4.2) 80(5.0) 95(5.0) 70(9.0) 50(5.0) 95(2.5) 54(1.8) 120(5.0) 65(3.2) 125(5.0) 90(2.5) 100(2.5) 120(3.0) 75(4.2) 80(2.5) 60(7.0) 50(9.0) 65(5.0) 75(5.0) 90(7.0) 90(9.0) 100(7.0) 50(7.0) 60(9.0) 70(7.0) 100(3.0)
Glass tubings, DURAN® borosilicate glass
Tubing Transfer Tubing
Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
  • High transmittance
  • Electrically highly insulating
  • High chemical resistance
  • High hydrolytic resistance
  • High resistance to thermal shock
  • High temperature resistance
Tube length ~1500 mm.