Supelco® Organic Reference Standard, Theaflavin

Supplier: SUPELCO

55016-1MG
55016-1MGEA 726 EUR
55016-1MG
Supelco® Organic Reference Standard, Theaflavin
Standards Organic Standards
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9252f3ffb7460fbc43fed0475a4e67891d823ab1&vid=9252f3ffb7460fbc43fed0475a4e67891d823ab1&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) 3,4,5-Trihydroxy-1,8-bis[(2R,3R)-3,5,7-trihydroxy-2-chromanyl]-6-benzo[7]annulenone; 3,4,6-Trihydroxy-1,8-bis(3α,5,7-trihydroxy-2α-chromanyl)-5H-benzocyclohepten-5-one
Environmentally Preferable
CAS No. 4670-05-7
MDL No. MFCD03427500
Molecular weight 564,49
Shipping conditions STD
Storage conditions –20 °C
M-Clarity quality level MQ100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR