UHPLC columns, Hypersil GOLD™ C18
Supplier: Thermo Fisher Scientific
25002-202130 25002-152130 25002-103030 25002-102130 25002-054630 25002-053030 25002-052130 25002-051030 25002-032130 25002-031030 25002-101030 25002-022130
554-4286EA 1290 EUR
554-4286 554-4285 554-4378 554-4284 554-4283 554-4377 554-4282 554-4642 554-4376 554-4640 554-4471 554-4638
UHPLC columns, Hypersil GOLD™ C18
Chromatography Columns
Endcapped, ultra-pure, silica-based columns with exceptional peak shape and resolution for HPLC and LC/MS.
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitivity
  • Confidence in the accuracy and quality of analytical data