Cation exchange HPLC columns, ProPac™ SCX-10 and WCX-10
Supplier: Thermo Fisher Scientific
063456 063462 074600 088766 088767 088768 088769 088779 063472 063474 063480 063700 054993 054994 054995
555-7289EA 1450.29 EUR
555-7289 555-7290 554-5300 555-7441 555-7442 555-7443 555-7444 555-7454 554-5302 554-5303 554-5304 555-7301 554-5301 554-5305 555-7274
Cation exchange HPLC columns, ProPac™ SCX-10 and WCX-10
Chromatography Columns
The ProPac HPLC columns are weak and strong cation exchange columns with exceptionally high resolution and efficiency for separations of protein variants.
  • Characterisation and quality control assessment of monoclonal antibodies and other proteins
  • Useful for characterisation of related protein variants including deamidation and monoclonal antibodies lysine truncation variants
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer graftedMore Product Information