HPLC columns, Acclaim™ C30
Supplier: Thermo Fisher Scientific
075718 075719 075720 075721 075722 075723 075724 075725 075726 078660 078661 078662 078664 078665 078666
554-5286EA 1090 EUR
554-5286 554-5287 554-5292 554-5293 554-5294 554-5288 554-5290 554-5291 554-5289 555-7562 555-7563 555-7564 555-7566 555-7567 555-7568
HPLC columns, Acclaim™ C30
Chromatography Columns
Acclaim™ C30 columns used to separate the structural isomers and unique complementary to other reversed-phase columns.
  • High shape selectively
  • Compatibility with highly aqueous mobile phase
  • High quality
  • Low column bleed
Order Now
SPECIFICATIONS
Environmentally Preferable