Phytochemical standard, Orientin, analytical standard, Supelco®
55736-5MG
55736-5MGEA 574 EUR
55736-5MG
Phytochemical standard, Orientin, analytical standard, Supelco®
Standards Organic Standards
Flavonoid glycoside found in the acai tree and globe and passion flowers. Antiviral, antimicrobial and antioxidant. Exhibits radioprotective activities.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56c6379e1e3155ca973af38a91f576e55108b8e2&vid=56c6379e1e3155ca973af38a91f576e55108b8e2&prid=web&clid=SAPDAM