Wetzel Waterjet pumps
Supplier: Lenz Laborglas GmbH & CO.KG
05585000 05585200
5585000.EA 120 EUR
5585000. 5585200.
Wetzel Waterjet pumps
Pumps Vacuum Water Jet Pumps
Water jet pumps made of borosilicate glass with or without non-return valve.
Order Now
SPECIFICATIONS
Environmentally Preferable