Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Warning


Management of Change (MOC) category = R

5683-07EA 1069.16 EUR
5683-07 5683-08 5683-09
Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®
Orthophosphoric acid
Formula: H₃PO₄
Density: 1,05 g/cm³
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Alkali Phosphates Passes Test
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Assay (as H₃PO₄), g/100 mL 9.5 - 10.5
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤ 6 ppm
EP/BP - Phosphorous or Hypophosphorous Acid Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Iron (Fe) ≤ 6 ppm
EP/BP - Substances Precipitated with Ammonia Passes Test
EP/BP - Sulfate (SO₄) ≤ 10 ppm
EP/BP - Assay (H₃PO₄) 9.5 - 10.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR