Sulphuric acid 95 - 98% 95-98%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Danger

6027.1000EA 79 EUR
6027.1000 6027.2500
Sulphuric acid 95 - 98% 95-98%, BAKER ANALYZED® ACS, J.T.Baker®
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay 95.0 - 98.0 %
Ammonium (NH4) max. 1 ppm
Appearance passes test
Color (APHA) max. 10
Mercury (Hg) max. 5 ppb
Residue after Ignition max. 4 ppm
Substances Reducing KMnO4(as SO2) max. 2 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.01
Aluminium (Al) max. 0.05
Arsenic (As) max. 0.01
Barium (Ba) max. 0.02
Beryllium (Be) max. 0.01
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.02
Chloride (Cl) max. 0.1
Cobalt (Co) max. 0.01
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.01
Iron (Fe) max. 0.1
Germanium (Ge) max. 0.05
Potassium (K) max. 0.1
Lithium (Li) max. 0.01
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.01
Molybdenum (Mo) max. 0.02
Sodium (Na) max. 0.5
Nickel (Ni) max. 0.05
Lead (Pb) max. 0.05
Strontium (Sr) max. 0.01
Titanium (Ti) max. 0.1
Thallium (Tl) max. 0.05
Vanadium (V) max. 0.01
Zinc (Zn) max. 0.05
Zirconium (Zr) max. 0.1
Heavy Metals (as Pb) max. 1
Nitrate (NO3) max. 0.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR