Digital stopwatch, 898M
Supplier: EA COMBS
898M
609-0006EA 207 EUR
609-0006
Digital stopwatch, 898M
Timers Digital Stopwatches
Stopwatch with water resistant case.
  • Timing capacity: 10 hours
  • Measuring range: ¹/₁₀₀ seconds
  • Times 99 laps and splits with counter indication
  • Recalls the first 100 splits and laps in memory
  • Stroke and frequency measurement based on 3 to 60
  • Time of day with calendar
Delivery information: Supplied with neck cord and CR2032 lithium battery for up to five years life under normal conditions.
Order Now
SPECIFICATIONS
Environmentally Preferable