VWR®, Safety Bottle Containers
Supplier: VWR Collection
611-1382EA 0 EUR
611-1382 611-1384
VWR®, Safety Bottle Containers
Carriers Bottle Carriers/Containers
These high tensile strength transport carriers are designed to ensure the safe transport of bottles containing acids, alkalis and solvents.
Rubber, coloured.
  • Handle specially shaped for carrying comfort
  • Service temperature from −45 to +110 °C; steam sterilisable
  • Locating rings in the bottom of the container ensure a gap of about 25 mm between the flask and the container wall
Small carriers
Hold bottles with a capacity of 0,5 or 1 litre.
Large carriers
Hold bottles with a capacity of 2,5 or 4 litres.
Order Now
SPECIFICATIONS
Environmentally Preferable