Precision balances, Pioneer PX series
Supplier: OHAUS
30429812 30429805 30430015 30429814 30429807 30430021 30429816 30429809 30430024 30619382 30619383 30619384 30619385 30714491 30619386 30868072 30868073
611-3842EA 1290 EUR
611-3842 611-3843 611-3844 611-3848 611-3849 611-3850 611-3854 611-3855 611-3856 611-5039 611-5040 611-5042 611-5044 611-5093 611-5091 OHAU30868072 OHAU30868073
Precision balances, Pioneer PX series
Balances and Scales
The Pioneer PX series combines essential weighing functionality with performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. PX models are economically priced, and intuitively designed for intelligent operation. Their durable design has a cast metal lower housing, metal sub-pan and stainless steel weighing pan for long-term use.
  • Large viewing angle with bright backlit LCD
  • 2-line display provides additional information or guidance prompts to user
  • A static removal bar (permanent static electric
More Product Information