Semi-micro balances, Pioneer PX® series
Supplier: OHAUS
30478196 30478197 30480168 30480169 30485340 30485341
611-4369EA 4160 EUR
611-4369 611-4370 611-4396 611-4397 611-4398 611-4399
Semi-micro balances, Pioneer PX® series
Balances and Scales
The Pioneer PX series of semi-micro balances combines essential weighing functionality with performance, offering high accuracy and repeatability for basic applications in laboratory, industrial and education settings. PX models are economically priced, and intuitively designed for intelligent operation. Their durable design has a cast metal lower housing, metal sub-pan and stainless steel weighing pan for long-term use.
  • Large viewing angle with bright backlit LCD
  • 2-line display provides additional information or guidance prompts to user
  • A static removal
More Product Information