Measuring cylinders, tall form, class A, borosilicate glass, with certificate, BLAUBRAND®
Supplier: Brand
2.32162 2.32164 2.32105 2.32108 2.32148 2.32128 2.32154 2.32138 2.32120
612-4615EA 98.3 EUR
612-4615 BRND2.32164 BRND2.32105 BRND2.32108 BRND2.32148 BRND2.32128 BRND2.32154 BRND2.32138 BRND2.32120
Measuring cylinders, tall form, class A, borosilicate glass, with certificate, BLAUBRAND®
Cylinders Measuring Cylinders
Borosilicate glass 3.3, tall form, class A, with individual certificate.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
Certifications: DIN EN ISO 4788
Order Now
SPECIFICATIONS
Environmentally Preferable