ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: EPPENDORF
0030077.598 0030078.624 0030078.586 0030078.500 0030078.519 0030078.527 0030078.535 0030078.543 0030078.551 0030078.560 0030078.578 0030078.594 0030078.616 0030078.810 0030078.829 0030078.837 0030078.845
613-3689EA 89.5 EUR
613-3689 613-6784 613-6781 613-6773 613-6774 613-6775 613-6776 613-6777 613-6778 613-6779 613-6780 613-6782 613-6783 613-6803 613-6804 613-6805 613-6806
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Pipette Tips
Eppendorf ep Dualfilter T.I.P.S.® Racks offer 'PCR clean and Sterile' grade filter pipette tips, providing dual-layer protection against contamination for critical lab applications, ensuring precision, reliability, and hygiene in high-stakes environments.
  • Two filter layers provide dual protection against aerosols and biomolecules to guard both pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent
More Product Information