VWR®, Environmentally Friendly Thermometers, Easy Read
Supplier: VWR Collection
620-0781EA 22.3 EUR
620-0781 620-0782 620-0784 620-0785 620-0786 620-0787 620-0788 620-0789
VWR®, Environmentally Friendly Thermometers, Easy Read
Thermometers Analogue Thermometers
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Eliminates reading errors
  • Non toxic, non hazardous, biodegradable liquid
Traceable to NIST/DKD. Compliant with Envirokleen™.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independentMore Product Information