VWR®, Environmentally Friendly Thermometers, Individually Calibrated, Easy Read
Supplier: VWR Collection
620-0828EA 495 EUR
620-0828 620-0830 620-0934 620-0935
VWR®, Environmentally Friendly Thermometers, Individually Calibrated, Easy Read
Thermometers Analogue Thermometers
Each easy to read thermometer includes an individualised thermometer calibration report stating corrections to one-tenth of the smallest division, unique report number, and NIST standards used for calibration. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • EnviroKleen™ certified
  • Non toxic, enviro safe liquid, against lead-free, yellow back glass provides easy readability and fewer reading errors