VWR®, Analogue Thermo-Hygrometer
Supplier: VWR Collection
620-0923EA 89.3 EUR
620-0923
VWR®, Analogue Thermo-Hygrometer
Hygrometers Analogue Hygrometers
For all environments, this analogue thermo-hygrometer can withstand use inside incubators, refrigerators and freezers. It features contrasting black temperature dial and red humidity dial against a white background.
  • Round design
  • Stainless steel casing
  • Large easy to read dial
Ø: 127 mm
Delivery information: Supplied with a hook for wall mounting.