VWR® Traceable®, Highly Accurate Freezer Thermometers
Supplier: VWR Collection
620-1976EA 223 EUR
620-1976 620-1977 620-1978 620-1979
VWR® Traceable®, Highly Accurate Freezer Thermometers
Thermometers Digital Thermometers
These triple-display thermometers offer a choice of different probes. The bottle insulates the thermometer from rapid temperature changes when door is opened. The stainless steel handle probe (Ø×L: 3,2×300 mm) with 3 metre cable is waterproof and suitable for liquids, air/gas and semi-solids, with adjustable holder and VELCRO® tape.
  • Maximum/minimum values on the last 24 hours (each hour) and the last 7 or 30 days (each day)
  • Time and date stamp key: Time and date for all minimum and maximum readings
  • Visual LED flashes and audible alarm sounds continuously
More Product Information