VWR® Traceable® excursion trac, Liquid Nitrogen USB Data Logging Thermometers
Supplier: VWR Collection
620-2633EA 589 EUR
620-2633 620-2634
VWR® Traceable® excursion trac, Liquid Nitrogen USB Data Logging Thermometers
Thermometers Digital Thermometers
These USB Traceable® excursion trac LN₂ data logging thermometers monitor the temperature in liquid nitrogen Dewars, covering cryogenic needs for sensitive, irreplaceable biological samples.
  • Memory capacity of 525 500 temperature observations
  • Four calibration points: −196; −80, 0 and 100 °C
  • Monitor readings overnight, at the weekend, or for any time period with rolling data log
  • User-defined data logging interval of 1 observation/minute to 1 observation/24 hours
  • Memory capacity maintains a year of recorded data when using 1 minute intervals and longer when
More Product Information