USB camera, Moticam® S1
Supplier: MOTIC
1100600101081
630-3099EA 549 EUR
630-3099
USB camera, Moticam® S1
Cameras
High Grade sCMOS USB camera Moticam® S1 for microscopy for an ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors with back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
Moticam S1 is an the entry model for back-illuminated sensor technology and speedy data transfer (with frame rates up to 120 fps*), with easy image setting and capturing. Create your perfect images for a professional database.
Ordering information: To be purchased separately – C-mount adapter not included, this needs to the same brand as yourMore Product Information