Microscope slides
Supplier: MARIENFELD
1000000 1000200 1000412 1000612 1120250
631-9460EA 3.2 EUR
631-9460 631-9461 631-9462 631-0015 MARI1120250
Microscope slides
Slides Microscope Slides
Slides designed for in vitro diagnostic (IVD) applications.
  • Pre-washed and ready to use
  • Either ground or cut versions
  • Soda-lime glass, Hydrolytic Class 3
Extra-white slides are used for low auto-fluorescence applications.