VWR®, Absorption Cuvettes
Supplier: VWR Collection
634-1196EA 103 EUR
634-1196 634-1197 634-1198 634-1199 634-1200 634-1201 634-1202 634-1203 634-1204 634-1205 634-1206 634-1207 634-1208 634-1209 634-1210 634-1211 634-1212 634-1213 634-1214 634-1215 634-1216 634-1217 634-1218 634-1219 634-1220 634-1221 634-1222
VWR®, Absorption Cuvettes
Cuvettes Spectrophotometer Cuvettes
High quality cuvettes manufactured from quartz or optical glass for accurate spectrophotometric analysis.
  • Fully heat-fused construction
  • Two cover types available: Flat lid and stopper
  • Matched sets available
Due to the fully heat-fused construction (no intermediate materials or adhesives), and careful annealing to remove any strain, there is no optical or physical distortion of the two optically polished windows. Flat lids and stoppers are made of polytetrafluoroethylene (PTFE). Stoppered cells provide a more suitable seal for volatile liquids. MatchedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable