Mesitylene, Supelco®

Supplier: SUPELCO
Danger


MQ100

63908-5ML
63908-5MLEA 240 EUR
63908-5ML
Mesitylene, Supelco®
Mesitylene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 165
Melting Pt: –45 °C
Density: 0.8637 g/cm³ (20 °C)
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00008538
CAS Number: 108-67-8
EINECS: 203-604-4
UN: 2325
ADR: 3 III
Merck Index: 13,05934
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d0b38dd9cab88abce28452647aada27e3f986df&vid=4d0b38dd9cab88abce28452647aada27e3f986df&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR