Titrator, Karl Fischer, volumetric, TitraLab® KF1000
Supplier: Hach
KF1121.82 KF1121.98
661-0222EA 5520 EUR
661-0222 661-0203
Titrator, Karl Fischer, volumetric, TitraLab® KF1000
Titrators
The TitraLab® KF1000 from Hach enables users to easily and quickly perform moisture content determinations. The automatic system requires only small sample quantities to deliver accurate and precise results for water content applications. The KF1000 utilises the application specific kit to make it quick and easy for anyone to set up and operate a test.
  • Eliminates operator interpretation and manual processes to quickly deliver accurate and repeatable results
  • Pre-programmed titration methods detect end points and eliminate manual calculations
  • Includes two
More Product Information